Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Caspase-3 Inhibitor, Z-DEVD-FMK

Catalog No. BDB550378 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550378 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550378 Supplier BD Supplier No. 550378

Irreversible and cell permeable inhibitor of caspase-3

Members of the caspase family play key roles in inflammation and mammalian apoptosis. The Z-DEVD-FMK peptide is O-methylated in the P1 position on aspartic acid providing enhanced stability and increased cell permeability. The Z-DEVD-FMK inhibitor is designed to be used in both in vivo and in vitro cell based assays to inhibit caspase-3 activity and to study events downsteam of caspase-3 activation.

  • Contents: Lyophilized; Reconstitute in DMSO
  • Format: Purified
  • Molecular weight: 668 Dalton
  • Storage buffer: Lyophilized in dimethyl sulfoxide (DMSO)

Avoid multiple freeze-thaws of product.

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Flow cytometry
Storage Buffer Lyophilized in Dimethyl Sulfoxide
Content And Storage -20°C
Quantity 1 mg
Molecular Weight (g/mol) 668
Inhibitors Z-DEVD-FMK
Form Purified

For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.