Learn More
Description
The CD1d:Ig fusion protein consists of extracellular major histocompatibility complex (MHC) class I CD1d domains that are fused with VH regions of mouse IgG1. Like MHC class I molecules, the CD1d noncovalently associates with β2 Microglobulin ( β2M). Recombinant CD1d molecules are useful for studying Natural Killer T (NKT)-cell function by immunofluorescent staining and flow cytometric analysis of antigen-specific NKT cells.
- Format: Purified
- Clone: DimerX/ CD1d
- Isotype: Mouse IgG1, λ
- Concentration: 0.5mg/mL
- Storage Buffer: Aqueous buffered solution containing ≤0.09% sodium azide
Specifications
Specifications
| Concentration | 0.5mg/mL |
| For Use With (Application) | ELISA, Flow Cytometry |
| Formulation | Aqueous Buffered solution containing ≤0.09% Sodium Azide |
| Name | Mouse CD1d:Ig Recombinant Fusion Protein |
| pH Range | 7.2 |
| Purification Method | Affinity Chromatography, SDS-PAGE |
| Quantity | 0.25 mg |
| Storage Requirements | 4°C |
| Cross Reactivity | Mouse |
| Recombinant | Recombinant |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.