Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cell Detachment Solution

Catalog No. bdb561527 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561527 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB561527 Supplier BD Supplier No. 561527
Only null left
Add to Cart
Add to Cart

Replacement for trypsin solution and can be used for the passaging of cell lines

  • Does not contain mammalian or bacterial derived products
  • Comprised of collagenolytic and proteolytic enzymes
  • Performs well when detaching cells for the analysis of many cell surface markers using flow cytometry and for cell sorting

Flow Cytometry, Cell Culture

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Product Type Cell Culture Dissociation Reagent
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.