Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytofix/Cytoperm™ Fixation/Permeabilization Solution Kit

Catalog No. bdb554714 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
250 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554714 250 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554714 Supplier BD Supplier No. 554714
Add to Cart
Add to Cart

Includes

  • Fixation/Permeabilization solution (125mL)
  • Perm/Wash Buffer, 10x concentrate containing Fetal Bovine Serum (FBS) and saponin (100mL)

Enables the fixation and permeabilization of cells which is necessary for staining intracellular cytokines with fluorochrome-conjugated anti-cytokine antibodies

  • Provides two reagents, fixation/permeabilization solution and Perm/Wash Buffer
  • After cell fixation and permeabilization, the Perm/Wash Buffer is used to wash the cells and to dilute the anti-cytokine antibodies for staining

TRUSTED_SUSTAINABILITY

Specifications

Quantity 250 Tests
Product Type Fixation/Permeabilization Reagent
Includes 125 mL fixation/permeabilization solution, perm/Wash™ buffer, fetal bovine serum (FBS) 10X and 100 mL saponin
No. of Tests 250
Regulatory Status RUO
Content And Storage 4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.