Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytofix™ Fixation Buffer

Catalog No. bdb554655 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554655 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554655 Supplier BD Supplier No. 554655
Add to Cart
Add to Cart

Can be used to fix unstained cells for subsequent immunofluorescent staining of intracellular cytokines. Can also be used to fix cells after immunofluorescent staining in order to preserve the light-scattering signals and fluorescent intensities of cells for analysis at a later time.

  • Intended to preserve human and rodent lymphoid cells for subsequent immunofluorescent staining of intracellular cytokines
  • Used to preserve light-scattering characteristics and fluorescence intensities of human and rodent hematopoietic cells that have been stained by immunofluorescence for subsequent flow cytometric analysis 
TRUSTED_SUSTAINABILITY

Specifications

Formulation Neutral pH Buffered Saline (PBS) with 4.21% Formaldehyde (w/w)
For Use With (Application) Can be used to fix unstained cells for subsequent immunofluorescent staining of intracellular cytokines
Content And Storage 4°C
Product Type Fixation Buffer
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.