Learn More
Description
The Functional Beads can be used in conjunction with the BD CBA Functional Bead Conjugation Buffer Set (Mfr. No. 558556) to covalently link water-soluble proteins to the surface of the beads via a sulfo-SMCC chemistry. The beads, once linked to a protein, can be used as a capture matrix for analytes of interest.
Specifications
Specifications
| Certifications/Compliance | RUO |
| Content And Storage | 4°C |
| Contents | Aqueous buffered solution containing ≤0.09% sodium azide |
| Description | Functional Bead (E4) |
| For Use With (Application) | Flow Cytometry |
| Material | Polystyrene |
| Product Type | Functional Bead (E4) |
| For Use With (Equipment) | Flow cytometer |
| No. of Tests | 6000 |
| Volume (Metric) | 0.9 mL |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.