Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Mouse/Rat Soluble Protein Master Buffer Kit, 500 tests

Catalog No. bdb558267 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
500 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558267 500 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558267 Supplier BD Supplier No. 558267
Only null left
Add to Cart
Add to Cart

Includes

Assay Diluent, Capture Bead Diluent, Detection Reagent Diluent, Wash Buffer, Instrument Setup Beads (A1, A9, F1, F9), PE Instrument Setup Bead F1, PE Positive Control Detector

Employs particles with discrete fluorescence intensities to detect soluble analytes at very low concentrations

This kit contains all supporting reagents necessary to perform an assay using BD™ CBA Mouse or Rat Soluble Protein Flex Set. Buffers and instrument setup reagents are optimized for analysis of analytes in tissue culture supernatants, plasma and serum samples.

  • Analyte working assay range: 10 to 2500pg/mL (mouse), 40 to 10000pg/mL (rat)
  • Should not be used with any non-Mouse or Rat Soluble Protein Flex Sets
  • Available in pack sizes of 100 and 500 tests

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
For Use With (Application) For analysis of analytes in tissue culture supernatants, plasma and serum samples
For Use With (Equipment) Flow cytometer
No. of Tests 500
Includes 1 bottle 150 mL assay diluent, 1 bottle 30 mL capture bead diluent, 1 bottle 30 mL detection reagent diluent, 1 bottle 650 mL wash buffer, 1 vial 0.25 mL instrument setup bead A1, 1 vial 0.25 mL instrument setup bead A9, 1 vial 1.0 mL instrument setup bead F1
Quantity 500 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.