Learn More
Description
This product contains lyophilized recombinant human Interferon-λ (IFN-λ)
- Storage Buffer: Lyophilized in aqueous buffered solution containing BSA and ProClin™ 150
Flow cytometry
Specifications
Specifications
| Product Type | Cytometric Bead Array (CBA) Standard |
| Description | Human IL-6 Flex Set Standard |
| No. of Tests | 100 |
| Content And Storage | 4°C |
| Quantity | 0.2 mL |
| Format | Lyophilized |
| Includes | Lyophilized recombinant Human Interferon-l (IFN-l) |
| Regulatory Status | RUO |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.