Learn More
Cytometric Bead Array Human Th1/Th2/Th17 Kit, BD
Catalog No. BDB560484 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
80 Tests
Catalog No. Quantity
BDB560484 80 Tests
1 options
Catalog No. BDB560484 Supplier BD Biosciences Supplier No. 560484
Add to Cart
Add to Cart
Includes
IL-2, IL-4, IL-6, IL-10, TNF, IFN-γ, IL-17A
Optimized for analysis of physiologically relevant concentrations (pg/mL levels) of specific cytokine proteins in tissue culture supernatants, EDTA plasma, and serum samples
Detection reagent provided in the kit is a mixture of phycoerythrin (PE)–conjugated antibodies, which provides a fluorescent signal in proportion to the amount of bound analyte.
  • Can be used to measure Interleukin-2 (IL-2), Interleukin-4 (IL-4), Interleukin-6 (IL-6), Interleukin-10 (IL-10), Tumor Necrosis Factor (TNF), Interferon-γ (IFN-γ), and Interleukin-17A (IL-17A) protein levels in a single sample
  • Provides sufficient reagents for 80 tests
For research use only. Not for use in diagnostic or therapeutic procedures.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2° to 8°C
No. of Tests 80.0
Includes 1 vial 0.8mL human IL-2 capture beads, 1 vial 0.8mL human IL-4 capture beads, 1 vial 0.8mL human IL-6 capture beads, 1 vial 0.8mL human IL-10 capture beads, 1 vial 0.8mL human TNF capture beads, 1 vial 0.8mL human IFN capture beads
Quantity 80 Tests
Validated Application RUO