Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytometric Bead Array Wash Buffer, BD

Catalog No. bdb560105 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
130mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560105 130mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560105 Supplier BD Biosciences Supplier No. 560105
Only null left
Add to Cart
Add to Cart

To be used with the BD CBA Flex Set assays

  • For wash steps and to resuspend beads for analysis

TRUSTED_SUSTAINABILITY

Specifications

Color White
Content And Storage 4°C
Format Liquid
Quantity 130mL
Validated Application 29 CFR 1910.1200, 91/155/EEC, 93/112/EC, RUO

For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.