Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Eosin Methylene Blue Agar, Levine
SCI_ED

Catalog No. S67106 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Eosin Methylene Blue Agar, Levine; 500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S67106 Eosin Methylene Blue Agar, Levine; 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S67106 Supplier BD Supplier No. 211221/EMD
This item is not returnable. View return policy

This dehydrated culture media is used for isolating and differentiating lactose-fermenting from lactose-nonfermenting gram-negative enteric bacilli.

Successful bacteriological work depends on good basics—and that includes high-quality agar and additives. So when you're planning your class projects, order ample supplies of these essential ingredients!
•    Agar acts as the solidifying agent in culture media
•    Additives can be used to enhance the growth of microorganisms

 

TRUSTED_SUSTAINABILITY

Specifications

Form Dehydrated
For Use With (Application) For gram negative enteric bacteria
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.