Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Dehydrated Media Endo Agar

Catalog No. B11199 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11199 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B11199 Supplier BD Supplier No. 211199
Add to Cart
Add to Cart
This item is not returnable. View return policy

Granuated media preserves homogeneity

  • Danger of inhalation reduced with dehydration

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage <30°C
Form Dehydrated
Product Type Agar
Formulation Dipotassium Phosphate 3.5g/L, Peptic digest of Animal Tissue 10g/L, Agar 15g/L, Lactose 10g/L, Sodium Sulfite 2.5g/L, Basic Fuchsin 0.5g/L
For Use With (Application) Detection of Coliform other enteric microorganisms
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.