Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Dehydrated Media Luria-Bertani Broth

Catalog No. B214906 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Content And Storage:
<30°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Content And Storage
B214906 <30°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B214906 Supplier BD Supplier No. 214906
Add to Cart
Add to Cart

For propagation and maintenance of E. coli for molecular biology

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage <30°C
Form Dehydrated
Product Type Microbiology Media
Formulation Tryptone 10g/L, Yeast extract 5g/L, Sodium Chloride 10g/L
For Use With (Application) For maintaining and propagating Escherichia coli in molecular microbiology procedures
Packaging Type Pail
Quantity 10,000 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.