Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Bacto™ Malt Extract Broth

Catalog No. B214912 Shop All Thermo Scientific Products
Change view
Click to view available options
Form:
Liquid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
B214912 Liquid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B214912 Supplier Gibco™ Supplier No. 214912

missing translation for 'validMainframeMsg'

For isolation and enumeration of molds and yeast

TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Product Type Microbiology Media
Formulation Malt extract 6g/L, Maltose, Technical 1.8g/L, Dextrose 6g/L, Yeast extract 1.2g/L
Packaging Type Pail
Quantity 10,000 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.