Learn More
BD Difco™ Antisera for Serological Confirmation of E. coli O157:H7
Shop All Waters Corporation ProductsDescription
E. coli O157:H7 is an enteric pathogen that requires only a low inoculum to cause disease. Transmission is usually via food items. Serotyping of the entero-hemorrhagic E. coli is useful in the epidemiological documentation of the spread of a particular strain in a foodborne outbreak. At the time of use, test both positive and negative control cultures to check performance of the antiserum.
- Product type: antigens and antisera
- Groupings: D through K Products
- Type: antisera
- Sub type: E.coli - Lyophilized, re-hydrate to 3mL
- Shelf Life: 1095 days
- Label has Natural Rubber Latex statement
Certifications
CE marked
Specifications
Specifications
| Certifications/Compliance | CE Marked |
| Content And Storage | Refrigerate, store at 2°C to 8°C |
| Product Type | E. Coli Antisera |
| For Use With (Application) | For tube test for serological confirmation of E. coli O157:H7 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.