Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Bordatella pertussis Serotyping Antisera

Catalog No. DF2309503 Shop All Waters Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF2309-50-3 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Contains Mercury. Dispose of properly.
Catalog No. DF2309-50-3 Supplier BD Supplier No. 223091
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Contains Mercury. Dispose of properly.

Rabbit serum for slide agglutination

  • Available in 1.0mL packaged vials

Certifications

CE marked

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE Marked
Content And Storage Refrigerate, store at 2°C to 8°C
For Use With (Application) Bordatella pertussis
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.