Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: 2x Yeast Extract Tryptone (2xYT) Medium

Catalog No. DF044017 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0440-17 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0440-17 Supplier BD Supplier No. 244020
Add to Cart
Add to Cart
This item is not returnable. View return policy

For cultivating recombinant strains of Escherichia coli. BD Difco™ Dehydrated Culture Media: 2x Yeast Extract Tryptone (2xYT) Medium is part of a wide selection of dehydrated culture media.

  • Formulation: Pancreatic digest of Casein 16g/L, Yeast extract 10g/L, Yeast extract 5g/L

Order Info

Prices are for units of one each unless otherwise specified.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage <30°C
Form Dehydrated
Media Type 2XYT
Product Type Microbiology Media
Formulation Pancreatic digest of Casein (Tryptone) 16g/L, Yeast extract 10g/L, NaCl 5g/L
For Use With (Application) Cultivation of recombinant strains of Escherichia coli
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.