Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Antibiotic Medium 3

Catalog No. DF0243178 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0243-17-8 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0243-17-8 Supplier BD Supplier No. 224320
Add to Cart
Add to Cart
This item is not returnable. View return policy

Penassay broth for determining antibiotic potency by the microbiological assay technique. BD Difco™ Dehydrated Culture Media: Antibiotic Medium 3 is part of a large selection of dehydrated media.

  • Penassay broth
  • Dehydrated
  • Culture Media
  • Antibiotic Medium 3
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance AOAC, EP, USP
Content And Storage <30°C
Form Dehydrated
Media Type Antibiotic Medium 3
Product Type Antibiotic Medium
Formulation Beef extract 1.5g/L, Yeast extract 1.5g/L, Peptone 5g/L, Dextrose 1g/L, Sodium Chloride 3.5g/L, Dipotassium Phosphate 3.68g/L, Monopotassium Phosphate 1.32g/L
For Use With (Application) Penassay broth
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.