Learn More
Description
- Penassay Seed Agar Microbiological assay of antibiotics
Specifications
Specifications
| Certifications/Compliance | EP, USP |
| Content And Storage | <30°C |
| Form | Dehydrated |
| Media Type | Antibiotic Medium 9 |
| Product Type | Agar |
| Formulation | Pancreatic digest of Casein 17g/L, Soy Peptone 3g/L, Dextrose 2.5g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 2.5g/L, Agar 20g/L |
| For Use With (Application) | Determination of antibiotic potency by the microbiological assay technique |
| Packaging Type | Bottle |
| Quantity | 500 g |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.