Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: APT Broth

Catalog No. DF0655179 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0655-17-9 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0655-17-9 Supplier BD Supplier No. 265510
Add to Cart
Add to Cart
This item is not returnable. View return policy

For cultivating heterofermentative lactobacilli and other organisms requiring high thiamine content

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance COMPF, USDA
Content And Storage 2°C to 8°C
Form Dehydrated
Media Type APT Agar
Product Type Microbiology Media
Formulation Yeast extract 7.5g/L, Pancreatic digest of Casein 12.5g/L, Dextrose 10g/L, Sodium Citrate 5g/L, Thiamine Hydrochloride 1mg/L, Sodium Chloride 5g/L, Dipotassium Phosphate 5g/L, Manganese Chloride 0.14g/L, Magnesium Sulfate 0.8g/L, Ferrous Sulfate 0.04g/L,
For Use With (Application) Cultivation of heterofermentative lactoBacilli and other organisms requiring high thiamine content
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.