Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: B12 Culture Agar

Catalog No. DF0541159 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0541-15-9 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0541-15-9 Supplier BD Supplier No. 254110
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For cultivating Lactobacillus delbrueckii subsp. lactis ATCC™ 7830 used in the Vitamin B12 Activity Assay

  • Formulation: Tomato Juice 5g/L, Proteose Peptone No.3 7.5g/L, Yeast extract 7.5g/L, Dextrose 10g/L, Monopotassium Phosphate 2g/L, Polysorbate 80 1g/L, Agar 14g/L

Certifications

USP

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance USP
Content And Storage <30°C
Form Dehydrated
Media Type B12 Culture Agar
Product Type Agar
Formulation Tomato Juice 5g/L, Proteose Peptone No.3 7.5g/L, Yeast extract 7.5g/L, Dextrose 10g/L, Monopotassium Phosphate 2g/L, Polysorbate 80 1g/L, Agar 14g/L
For Use With (Application) For Lactobacillus delbrueckii
Packaging Type Petri Dish
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.