Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: B12 Inoculum Broth

Catalog No. DF0542158 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0542-15-8 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0542-15-8 Supplier BD Supplier No. 254210
Add to Cart
Add to Cart
This item is not returnable. View return policy

For preparing the inoculum of L. delbrueckii subsp. lactis ATCC™ 7830 used in the Vitamin B12 Activity Assay

  • Formulation: Tomato Juice 5g/L, Proteose Peptone No.3 7.5g/L, Yeast extract 7.5g/L, Dextrose 10g/L, Dipotassium Phosphate 2g/L, Polysorbate 80 1g/L

Certifications

USP

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance USP
Content And Storage <30°C
Form Dehydrated
Media Type B12 Inoculum Broth
Product Type Microbiology Media
Formulation Tomato Juice 5g/L, Proteose Peptone No.3 7.5g/L, Yeast extract 7.5g/L, Dextrose 10g/L, Dipotassium Phosphate 2g/L, Polysorbate 80 1g/L
For Use With (Application) Cultivation of wide variety of microorganisms
Packaging Type Bottle
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.