Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Biotin Assay Medium

Catalog No. DF0419158 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Biotin Assay Medium; 100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
DF0419-15-8 Biotin Assay Medium; 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0419-15-8 Supplier BD Supplier No. 241910
This item is not returnable. View return policy

For determining biotin concentration by the microbiological assay technique

Certifications

ISO, 9000

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO, 9000
Content And Storage 2°C to 8°C
Form Dehydrated
Media Type Biotin Assay Medium
Product Type Biotin Assay Medium
Formulation Vitamin assay Casamino Acids 12g/L, Dextrose 40g/L, Sodium Acetate 20/L, L-Cystine 0.2g/L, DL-Tryptophan 0.2g/L, Adenine Sulfate 20mg/L, Guanine Hydrochloride 20mg/L, Uracil 20mg/L, Thiamine Hydrochloride 2mg/L, Riboflavin 2mg/L, Niacin 2mg/L, Calcium Pantothenate 2mg/L, Pyridoxine Hydrochloride 4mg/L, p-Aminobenzoic Acid 200μg/L, Dipotassium Phosphate 1g/L, Monopotassium Phosphate 1g/L, Magnesium Sulfate 0.4g/L, Sodium Chloride 20mg/L, Ferrous Sulfate 20mg/L, Manganese Sulfate 20mg/L
For Use With (Application) Isolation of Lactobacillus plantarum 8014
Packaging Type Bottle
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.