Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Brilliant Green Agar
GSA_VA

Catalog No. DF0285177 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Brilliant Green Agar; 500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
DF0285-17-7 Brilliant Green Agar; 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0285-17-7 Supplier BD Supplier No. 228530
Add to Cart
Add to Cart
This item is not returnable. View return policy

Dehydrated form for convenience of laboratorian and bioprocessing specialist

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, EP, SMWW
Content And Storage Refrigerate
Form Dehydrated
Media Type Brilliant Green Agar
Product Type Brilliant Green Agar
Formulation Proteose Peptone 10g/L, Yeast extract 3g/L, Lactose 10g/L, Saccharose 10g/L, Sodium Chloride 5g/L, Agar 20g/L, Brilliant green 12.5mg/L, Phenol red 0.08g/L
For Use With (Application) Isolation of Salmonella other than S. Typhi from feces and other material
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.