Learn More
BD Difco™ Dehydrated Culture Media: Buffered Listeria Enrichment Broth Base
Shop All BD Diagnostic Systems Products

Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Certifications/Compliance | BAM, COMPF, SMD |
| Content And Storage | Refrigerate |
| Form | Dehydrated |
| Media Type | Buffered Listeria Enrichment Broth |
| Product Type | Microbiology Media |
| Formulation | Pancreatic digest of Casein 17g/L, Soytone 3g/L, Dextrose 2.5g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 2.5g/L, Disodium Phosphate 9.6g/L, Monopotassium Phosphate 1.35g/L, Yeast extract 6g/L, Sodium Pyruvate 1.1g/L |
| For Use With (Application) | Isolation of Listeria |
| Packaging Type | Bottle |
| Quantity | 500 g |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.