Learn More
BD Difco™ Dehydrated Culture Media: Buffered Peptone Water Preenrichment
Shop All BD Diagnostic Systems ProductsDescription
Certifications
ISO/DIS, 11014, BAM, CCAM, ISO, USDA
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014, BAM, CCAM, ISO, USDA |
| Content And Storage | <30°C |
| Form | Dehydrated |
| Media Type | Peptone Water |
| Product Type | Dehydrated Culture Media |
| Formulation | Peptone 10g/L, Sodium Chloride 5g/L, Disodium Phosphate 3.5g/L, Monopotassium Phosphate 1.5g/L |
| For Use With (Application) | Isolation of Salmonella species |
| Packaging Type | Pail |
| Quantity | 2 kg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.