Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Columbia Blood Agar Base, Enhanced Haemolysis

Catalog No. DF0790175 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0790-17-5 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0790-17-5 Supplier BD Supplier No. 279030
Add to Cart
Add to Cart
This item is not returnable. View return policy

For enhanced beta-hemolytic reactions after overnight incubation and for cultivation of fastidious microorganisms

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance BS12, CMPH2, MCM9
Content And Storage <30°C
Form Dehydrated
Media Type Columbia Blood Agar Base
Product Type Agar
Formulation Pantone 12g/L, Bitone H Plus 6g/L, Enzymatic digest of Animal Tissue 3g/L, Starch 1g/L, Sodium Chloride 5g/L, Agar 12g/L
For Use With (Application) For enhanced beta-hemolytic reactions after overnight incubation and for cultivation of fastidious microorganisms
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.