Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Cooke Rose Bengal Agar

Catalog No. DF0703171 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0703-17-1 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0703-17-1 Supplier BD Supplier No. 270310
Add to Cart
Add to Cart
This item is not returnable. View return policy

Antimicrobic vial A for selective isolation of fungi from environmental and food specimens

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance SMWW
Content And Storage <30°C
Form Dehydrated
Media Type Cooke Rose Bengal Agar
Product Type Agar
Formulation Soy Peptone 5g/L, Dextrose 10g/L, Monopotassium Phosphate 1g/L, Magnesium Sulfate 0.5g/L, Agar 20g/L, Rose Bengal 35mg/L
For Use With (Application) Antimicrobic vial A for the selective isolation of fungi from environmental and food specimens
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.