Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Cystine Lactose Electrolyte Deficient (CLED) Agar

Catalog No. L4312218 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Cystine Lactose Electrolyte Deficient (CLED) agar; 500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
L4312218 Cystine Lactose Electrolyte Deficient (CLED) agar; 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. L4312218 Supplier BD Supplier No. 212218
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Isolation, enumeration and presumptive identification of microorganisms from urine

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Form Dehydrated
Media Type CLED Agar
Product Type Agar
Formulation Pancreatic digest of Gelatin 4g/L, Pancreatic digest of Casein 4g/L, Beef extract 3g/L, Lactose 10g/L, L-Cystine 128mg/L, Bromthymol blue 0.02g/L, Agar 15g/L
For Use With (Application) Isolation, enumeration and presumptive identification of microorganisms from urine
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.