Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Cystine Tryptic Agar

Catalog No. DF0523179 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0523-17-9 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0523-17-9 Supplier BD Supplier No. 252310
Add to Cart
Add to Cart
This item is not returnable. View return policy

For cultivation of fungi and bacteria capable of using inorganic Nitrogen

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance AOAC
Content And Storage <30°C
Form Dehydrated
Media Type Cystine Tryptic Agar
Product Type Agar
Formulation Tryptose 20g/L, L-Cystine 0.5g/L, Sodium Chloride 5g/L, Sodium Sulfite 0.5g/L, Agar 2.5g/L, Phenol red 17mg/L
For Use With (Application) Cultivation of fungi and bacteria capable of using inorganic Nitrogen
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.