Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: CzaPEK Solution Agar

Catalog No. df0339173 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0339-17-3 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0339-17-3 Supplier BD Supplier No. 233910
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

For cultivation of organisms capable of utilizing inorganic nitrogen. BD Difco™ Dehydrated Culture Media: CzaPEK Solution Agar is the perfect choice.

  • Microbiology media
  • Dehydrated
  • Solution Agar
  • 500g
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance SMWW
Content And Storage 2°C to 8°C
Form Dehydrated
Media Type CzaPEK Solution Agar
Product Type Agar
Formulation Saccharose 30g/L, Sodium Nitrate 2g/L, Dipotassium Phosphate 1g/L, Magnesium Sulfate 0.5g/L, Potassium Chloride 0.5g/L, Ferrous Sulfate 0.01g/L, Agar 15g/L
For Use With (Application) Cultivation of organisms capable of utilizing inorganic Nitrogen
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.