Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: D/E Neutralizing Agar (Disinfectant Testing)

Catalog No. DF0686172 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0686-17-2 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0686-17-2 Supplier BD Supplier No. 268620
Add to Cart
Add to Cart
This item is not returnable. View return policy

Tests efficiency of disinfectants and sanitation

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance COMPF, SMD
Content And Storage Refrigerate
Form Dehydrated
Media Type D/E Neutralizing Agar
Product Type Agar
Formulation Pancreatic digest of Casein 5g/L, Yeast extract 2.5g/L, Dextrose 10g/L, Sodium Thioglycollate 1g/L, Sodium Thiosulfate 6g/L, Sodium Bisulfite 2.5g/L, Polysorbate 80 5g/L, Lecithin 7g/L, Bromcresol Purple 0.02g/L, Agar 15g/L
For Use With (Application) Testing of disinfectants and sanitation efficiency
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.