Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Desoxycholate Agar

Catalog No. DF0273171 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0273-17-1 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0273-17-1 Supplier BD Supplier No. 227310
Add to Cart
Add to Cart
This item is not returnable. View return policy

For isolating and differentiating gram negative enteric Bacilli

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, COMPF, SMWW
Content And Storage Refrigerate
Form Dehydrated
Media Type Desoxycholate Agar
Product Type Agar
Formulation Peptone 10g/L, Lactose 10g/L, Sodium Desoxycholate 1g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 2g/L, Ferric Ammonium Citrate 1g/L, Sodium Citrate 1g/L, Agar 15g/L, Neutral red 0.03g/L
For Use With (Application) For isolating and differentiating gram negative enteric Bacilli
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.