Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Desoxycholate Lactose Agar

Catalog No. DF0420173 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0420-17-3 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0420-17-3 Supplier BD Supplier No. 242010
Add to Cart
Add to Cart
This item is not returnable. View return policy

For isolating and differentiating gram negative enteric bacilli, for enumerating Coliforms from Water, waste Water, Milk and dairy products

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Refrigerate
Form Dehydrated
Media Type Desoxycholate Lactose Agar
Product Type Agar
Formulation Proteose Peptone 10g/L, Lactose 10g/L, Sodium Desoxycholate 0.5g/L, Sodium Chloride 5g/L, Sodium Citrate 2g/L, Agar 15g/L, Neutral red 0.03g/L
For Use With (Application) For isolating and differentiating gram negative enteric bacilli, for enumerating Coliforms from Water, waste Water, Milk and dairy products
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.