Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: DNase Test Agar (Deoxyribonuclease Test)

Catalog No. DF0632177 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0632-17-7 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0632-17-7 Supplier BD Supplier No. 263220
Add to Cart
Add to Cart
This item is not returnable. View return policy

Differentiates microorganisms based on DNase activity

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance COMPF
Content And Storage Refrigerate
Form Dehydrated
Media Type DNase Test Agar
Product Type Agar
Formulation Tryptose 20g/L, Deoxyribonucleic Acid 2g/L, Sodium Chloride 5g/L, Agar 15g/L
For Use With (Application) For differentiating microorganisms based on DNase activity
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.