Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: DRBC Agar (Dichloran Rose Bengal Chloramphenicol)

Catalog No. DF058717 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0587-17 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0587-17 Supplier BD Supplier No. 258710
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Supports good growth of yeasts and molds

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance BAM, CCAM, COMPF, SMD
Content And Storage Refrigerate
Form Dehydrated
Media Type DRBC Agar
Product Type Agar
Formulation Proteose Peptone No. 3 5g/L, Dextrose 10g/L, Monopotassium Phosphate 1g/L, Magnesium Sulfate 0.5g/L, Dichloran 2mg/L, Rose Bengal 25mg/L, Chloramphenicol 0.1g/L, Agar 15g/L
For Use With (Application) Supports good growth of Yeasts and molds while inhibiting bacteria and the spreading of rapidly growing molds
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.