Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Dubos Broth Base

Catalog No. DF0385176 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0385-17-6 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0385-17-6 Supplier BD Supplier No. 238510
Add to Cart
Add to Cart
This item is not returnable. View return policy

Used with Dubos medium Albumin Cultivation of pure cultures of M. tuberculosis

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Form Dehydrated
Media Type Dubos Broth Base
Product Type Microbiology Media
Formulation Pancreatic digest of Casein 0.5g/L, Asparagine 2g/L, Polysorbate 80 0.2g/L, Monopotassium Phosphate 1g/L, Disodium Phosphate (anhydrous) 2.5g/L, Ferric Ammonium Citrate 50mg/L, Magnesium Sulfate 10mg/L, Calcium Chloride 0.5mg/L, Zinc Sulfate 0.1mg/L, Copper Sulfate 0.1mg/L
For Use With (Application) Used with Dubos medium Albumin Cultivation of pure cultures of M. tuberculosis
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.