Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Elliker Broth (LactoBacilli broth)

Catalog No. df0974173 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0974-17-3 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0974-17-3 Supplier BD Supplier No. 212183
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

For cultivating streptococci and lactobacilli, particularly in dairy procedures

  • Formulation: Pancreatic digest of Casein 20g/L, Yeast extract 5g/L, Gelatin 2.5g/L, Dextrose 5g/L, Lactose 5g/L, Saccharose 5g/L, Sodium Chloride 4g/L, Sodium Acetate 1.5g/L, Ascorbic Acid 0.5g/L

Certifications

SMD

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance SMD
Content And Storage <30°C
Form Dehydrated
Media Type Elliker Broth
Product Type Microbiology Media
Formulation Pancreatic digest of Casein 20g/L, Yeast extract 5g/L, Gelatin 2.5g/L, Dextrose 5g/L, Lactose 5g/L, Saccharose 5g/L, Sodium Chloride 4g/L, Sodium Acetate 1.5g/L, Ascorbic Acid 0.5g/L
For Use With (Application) Cultivation of lactoBacilli and streptococci
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.