Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Esculin Iron Agar

Catalog No. DF0488154 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0488-15-4 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0488-15-4 Supplier BD Supplier No. 248810
Add to Cart
Add to Cart
This item is not returnable. View return policy

For enumerating enterococci from water by membrane filtration based on esculin hydrolysis

  • Formulation: Esculin 1g/L, Ferric Ammonium Citrate 0.5g/L, Agar 15g/L

Certifications

EPa, SMWW

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EPa, SMWW
Content And Storage 2°C to 8°C
Form Dehydrated
Media Type EIA Medium
Product Type Agar
Formulation Esculin 1g/L, Ferric Ammonium Citrate 0.5g/L, Agar 15g/L
For Use With (Application) For enumerating Enterococci from Water by membrane filtration based on Esculin hydrolysis
Packaging Type Bottle
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.