Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Eugon Agar

Catalog No. DF0589170 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0589-17-0 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0589-17-0 Supplier BD Supplier No. 258910
Add to Cart
Add to Cart
This item is not returnable. View return policy

General-purpose medium for cultivating a wide variety of microorganisms

  • Formulation: Proteose Peptone 7.5g/L, Pancreatic digest of Casein 7.5g/L, Soy Peptone 5g/L, Dextrose 5.5g/L, L-Cystine 0.7g/L, Sodium Chloride 4g/L, Sodium Sulfite 0.2g/L, Agar 15g/L

Certifications

CE, COMPF

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, COMPF
Content And Storage <30°C
Form Dehydrated
Media Type Eugon Agar
Product Type Agar
Formulation Proteose Peptone 7.5g/L, Pancreatic digest of Casein 7.5g/L, Soy Peptone 5g/L, Dextrose 5.5g/L, L-Cystine 0.7g/L, Sodium Chloride 4g/L, Sodium Sulfite 0.2g/L, Agar 15g/L
For Use With (Application) Cultivation of a variety of organisms, well suited for large scale vaccine production
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.