Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Fluid Thioglycollate Medium with Beef Extract

Catalog No. DF0697179 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
10,000 g
500 g
Packaging Type:
Bottle
Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
DF0697-17-9 500 g Bottle
DF0697-08 10,000 g Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. DF0697-17-9 Supplier BD Supplier No. 269720
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used in cultivating microorganisms from normally sterile biological products

  • Recommended by the Agricultural Research Service
  • Formulation: Beef extract 5g/L, Yeast extract 5g/L, Pancreatic digest of Casein 15g/L, Dextrose 5.5g/L, Sodium Chloride 2.5g/L, L-Cystine 0.5g/L, Sodium Thioglycollate 0.5g/L, Agar 0.75g/L, Resazurin 1mg/L

Certifications

CE, USP

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, USP
Content And Storage Refrigerate
Form Dehydrated
Media Type Fluid Thioglycollate Medium
Product Type Dehydrated Culture Media
Formulation Beef extract 5g/L, Yeast extract 5g/L, Pancreatic digest of Casein 15g/L, Dextrose 5.5g/L, Sodium Chloride 2.5g/L, L-Cystine 0.5g/L, Sodium Thioglycollate 0.5g/L, Agar 0.75g/L, Resazurin 1mg/L
For Use With (Application) Cultivation of microorganisms from normally sterile biological product
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.