Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Folic Acid Assay Medium

Catalog No. DF0318150 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0318-15-0 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0318-15-0 Supplier BD Supplier No. 231810
Add to Cart
Add to Cart
This item is not returnable. View return policy

For determining folic acid concentration by the microbiological assay technique

  • Formulation: Vitamin assay Casamino Acids 12g/L, Dextrose 40g/L, Sodium Citrate 20g/L, L-Cystine 0.2g/L, DL-Tryptophan 0.2g/L, Adenine Sulfate 20mg/L, Guanine Hydrochloride 20mg/L, Uracil 20mg/L, Thiamine Hydrochloride 2mg/L, Pyridoxine Hydrochloride 4mg/L, Riboflavin, Niacin 2mg, Calcium Pantothenate 400μg, p-Aminobenzoic Acid 200μg, Biotin 0.8μg, Dipotassium Phosphate 1g, Monopotassium Phosphate 1g, Magnesium Sulfate 0.4g, Sodium Chloride 20mg, Ferrous Sulfate 20mg, Manganese Sulfate 20mg

Certifications

ISO, 9000

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO, 9000
Content And Storage 2°C to 8°C
Form Dehydrated
Media Type Folic Acid Assay Medium
Product Type Microbiology Media
Formulation Vitamin assay Casamino Acids 12g/L, Dextrose 40g/L, Sodium Citrate 20g/L, L-Cystine 0.2g/L, DL-Tryptophan 0.2g/L, Adenine Sulfate 20mg/L, Guanine Hydrochloride 20mg/L, Uracil 20mg/L, Thiamine Hydrochloride 2mg/L, Pyridoxine Hydrochloride 4mg/L, Riboflavin 2mg/L, Niacin 2mg/L, Calcium Pantothenate 400μg/L, p-Aminobenzoic Acid 200μg/L, Biotin 0.8μg/L, Dipotassium Phosphate 1g/L, Monopotassium Phosphate 1g/L, Magnesium Sulfate 0.4g/L, Sodium Chloride 20mg/L, Ferrous Sulfate 20mg/L, Manganese Sulfate 20mg/L
For Use With (Application) For determining folic acid concentration by the microbiological assay technique
Packaging Type Bottle
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.