Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Gonococcal (GC) Medium Base

Catalog No. DF0289173 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0289-17-3 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0289-17-3 Supplier BD Supplier No. 228950
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For various additives in isolating and cultivating Neisseria gonorrhoeae and other fastidious microorganisms. BD Difco™ Dehydrated Culture Media: Gonococcal (GC) Medium Base is part of a large selection of dehydrated media.

  • Dehydrated
  • Culture Media
  • Gonococcal (GC) Medium Base
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage <30°C
Form Dehydrated
Media Type GC Medium Base
Product Type Microbiology Media
Formulation Proteose Peptone No. 3 15g/L, Corn Starch 1g/L, Dipotassium Phosphate 4g/L, Monopotassium Phosphate 1g/L, Sodium Chloride 5g/L, Agar 10g/L
For Use With (Application) For various additives in isolating and cultivating Neisseria gonorrhoeae and other fastidious microorganisms
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.