Learn More
Specifications
Specifications
| Certifications/Compliance | CE |
| Content And Storage | Refrigerate |
| Form | Dehydrated |
| Media Type | ISP-Yeast Malt Agar (Medium 2) |
| Product Type | Agar |
| Formulation | Yeast extract 4g/L, Malt extract 10g/L, Dextrose 4g/L, Agar 20g/L |
| For Use With (Application) | For characterizing Streptomyces species according to the International Streptomyces project |
| Packaging Type | Bottle |
| Quantity | 500 g |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.