Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: ISP Medium 4

Catalog No. DF0772177 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0772-17-7 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0772-17-7 Supplier BD Supplier No. 277210
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

International Streptomyces Project Inorganic Salts Starch Agar

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE
Content And Storage Refrigerate
Form Dehydrated
Media Type ISP-Yeast Malt Agar (Medium 4)
Product Type Agar
Formulation Soluble Starch 10g/L, Dipotassium Phosphate 1g/L, Magnesium Sulfate USP 1g/L, Sodium Chloride 1g/L, Ammonium Sulfate 2g/L, Calcium Carbonate 2g/L, Ferrous Sulfate 1mg/L, Manganous Chloride 1mg/L, Zinc Sulfate 1mg/L, Agar 20g/L
For Use With (Application) For characterizing Streptomyces species according to the International Streptomyces project
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.