Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: KF Streptococcus Agar

Catalog No. DF0496172 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0496-17-2 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0496-17-2 Supplier BD Supplier No. 249610
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use with TTC or TTC Solution 1%, if desired for selective enumeration of fecal Streptococci

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE
Content And Storage Refrigerate
Form Dehydrated
Media Type KF Streptococcus Agar
Product Type Agar
Formulation Proteose Peptone 10g/L, Yeast extract 10g/L, Sodium Chloride 5g/L, Sodium Glycerophosphate 10g/L, Maltose 20g/L, Lactose 1g/L, Sodium Azide 0.4g/L, Bromcresol Purple 15mg/L, Agar 20g/L
For Use With (Application) Used with TTC or TTC solution 1%, if desired For selective enumeration of fecal streptococci
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.