Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: King Medium A (Pseudomonas agar P)

Catalog No. DF0449170 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0449-17-0 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0449-17-0 Supplier BD Supplier No. 244910
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Use with Glycerol for detection and differentiation of P. aeruginosa based on pyocyanin production

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, BAM
Content And Storage Refrigerate
Form Dehydrated
Media Type King Medium A
Product Type Microbiology Media
Formulation Pancreatic digest of Gelatin 20g/L, Magnesium Chloride 1.4g/L, Potassium Sulfate 10g/L, Agar 15g/L
For Use With (Application) Used with Glycerol for detection and differentiation of P. aeruginosa based on Pyocyanin production
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.