Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Lactobacilli Agar AOAC

Catalog No. DF0900154 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0900-15-4 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0900-15-4 Supplier BD Supplier No. 290010
Add to Cart
Add to Cart
This item is not returnable. View return policy

AOAC Maintenance of stock cultures for vitamin and amino acid microbiological assays

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance AOAC
Content And Storage 2°C to 8°C
Form Dehydrated
Media Type Lactobacilli Agar
Product Type Agar
Formulation Peptonized Milk 15g/L, Yeast extract 5g/L, Dextrose 10g/L, Tomato Juice 5g/L, Monopotassium Phosphate 2g/L, Polysorbate 80 1g/L, Agar 10g/L
For Use With (Application) AOAC maintenance of stock cultures for Vitamin and Amino Acid microbiological assays
Packaging Type Bottle
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.