Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Lactobacilli Broth AOAC

Catalog No. DF0901153 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0901-15-3 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0901-15-3 Supplier BD Supplier No. 290110
This item is not returnable. View return policy

AOAC Used to prepare microorganism inocula for vitamin assays

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance AOAC
Content And Storage 2°C to 8°C
Form Dehydrated
Media Type Lactobacilli Broth
Product Type Microbiology Media
Formulation Peptonized Milk 15g/L, Yeast extract 5g/L, Dextrose 10g/L, Tomato Juice 5g/L, Monopotassium Phosphate 2g/L, Polysorbate 80 1g/L
For Use With (Application) AOAC used to prepare microorganisms inocula for Vitamin assays
Packaging Type Bottle
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.