Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Lauryl Tryptose Broth with MUG

Catalog No. DF0021176 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0021-17-6 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0021-17-6 Supplier BD Supplier No. 211744
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Broth with MUG Fluorogenic detection of E. coli in water, food and milk

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, AOAC, BAM, COMPF, SMD, SMWW
Content And Storage <30°C
Form Dehydrated
Media Type Lauryl Tryptose Broth
Product Type Microbiology Media
Formulation Tryptose 20g/L, Lactose 5g/L, Dipotassium Phosphate 2.75g/L, Monopotassium Phosphate 2.75g/L, Sodium Chloride 5g/L, Sodium Lauryl Sulfate 0.1g/L, 4-methylumbelliferyl-beta-D-glucuronide0.05g/L
For Use With (Application) Detection of Escherichia coli in Water, food and dairy samples by a fluorogenic procedure
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.